Would you like to log in again?
Thermolabile Proteinase K
Supplier: New England Biolabs
P8111S
76326-052EA 177 USD InStock
76326-052
Thermolabile Proteinase K
Enzymes
Thermolabile Proteinase K (TLPK) is an engineered, subtilisin-related serine protease that will hydrolyze a broad variety of peptide bonds.
  • Optimal activity and stability for up to 24 months
  • Active in a wide range of reaction buffers with optimal activity between 20 to 40 °C and pH 7.0 to 9.5
  • No detectable endonuclease, exonuclease, DNase or RNase contaminating activities
It preferentially cleaves the peptide bond at the carboxyl side of aliphatic or aromatic amino acid residues. TLPK can be completely inactivated by incubation at 55 °C for 10 minutes, which allows for subsequent enzymatic steps in the same reaction vessel.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty