Would you like to log in again?
BioClean™ Cut Resistant Glove Liner, S-BCRL, Ansell
Supplier: Ansell
BioClean™
S-BCRL-5055 S-BCRL-8085 S-BCRL-7075 S-BCRL-6065 S-BCRL-9090
76326-070CS 2760.52 USD InStock
76326-070 76326-064 76326-066 76326-068 76326-144
BioClean™ Cut Resistant Glove Liner, S-BCRL, Ansell
Gloves
BioClean™ sterile cut-resistant glove liners feature Dyneema® Diamond yarn and provide cut resistance and protection during rigorous procedures.
  • Optimal dexterity, comfort, and fit
  • Recommended to be worn between two cleanroom gloves
  • Ultra high molecular weight polyethylene material
  • Supplied sterile via gamma irradiation
  • Latex-free, contains no NRL proteins
These cut-resistant glove liners are designed to be worn between two cleanroom gloves to provide EN 388 Level B and ANSI A2 cut protection when handling sharp objects or cleaning apparatus which pose a cut risk. Ideally suited for use in life science, chemical, and healthcare industries.
Glove liners can be autoclaved for up to 5 cycles at 121 °C for 30 minutes.
Certifications: Mechanical protection EN388:2016 Level B and ANSI A2. Glove Standard EN 420:2003+A1:2009.
Packaging: Glove liners are packaged one pair per inner PE wallet; one wallet per sealed EasyTear PE pouch; 10 pouches per sealed outer PE bag; 10 outer bags per lined carton.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty