Would you like to log in again?
HEAT GUN MASTER 800DEGF 230V EURO PLUG
Supplier: MASTER APPLIANCE
Heat Gun Master 800Degf 230V Euro Plug^ Master* Heat gun, long-life universal motor, high 27 CFM airflow which gets tough jobs done faster, a quick-change plug-in heating element, designed for serviceability, ergonomic, soft grip handle that maximizes user comfort, 800degF 230V EURO PLUG
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HEAT GUN MASTER 800DEGF 230V EURO PLUG 76326-106 HG-302D-03 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty