Would you like to log in again?
Fructose-1,6-Bisphosphatase Activity Assay Kit (Colorimetric), BioVision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AB273329-100TEST
76326-204EA 880.01 USD InStock
76326-204
Fructose-1,6-Bisphosphatase Activity Assay Kit (Colorimetric), BioVision
Assays Enzyme Assays
First assay in the market that measures FBP activity in biological samples.
  • Sensitive, specific assay
  • Validated with animal and cell culture lysates
  • Includes FBP positive control
  • Measures FBP enzymatic activity in biological assays
Fructose-1,6-Bisphosphatase (FBP) (EC 3.1.3.11) is a rate-limiting enzyme for gluconeogenesis. It hydrolyzes Fructose-1,6-Bisphosphate releasing inorganic phosphate and Fructose-6-Phosphate that is further used in the synthesis of glucose. In humans, Fructose-1,6-Bisphosphatase can be found in the form of two isozymes: FBP1 and FBP2. FBP1 is mainly expressed in liver and kidneys, while FBP2 is expressed in all tissues. Deficiency of Fructose-1,6-Bisphoshatase causes hypoglycemia, metabolic acidosis, and unexpected death in infants. Recent studies have found elevated FBP levels subjects dealing with obesity and diabetes. Furthermore, inhibition of FBP increases glucose utilization and insulin sensitivity. Therefore, FBP could serve as a potential target for treating type II diabetes (insulin resistance). BioVision’s FBP activity assay kit provides a quick, reliable test that allows the measurement of FBP enzymatic activity in various samples. In this assay, FBP hydrolyzes Fructose-1,6-Bisphosphate into Fructose-6-Phosphate (F6P). F6P is used in an enzyme-coupled reaction which reduces a chromophore forming a product with a stable signal that can measured at OD = 450 nm. The assay is simple, sensitive and can detect lower than 100 µU of FBP in variety of samples.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty