Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Megafuge™ ST4 Plus-MD Centrifuges, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

75009504 75016050CAP 75016037 75016049 75016050 75016037CAP 75016070 75009504CAP 75016049CAP 75016038 75009618 75009519 75009503 75016038CAP 75009518 75016069 75009519CAP 75016060
76326-240EA 12896.84 USD InStock
76326-240 76326-284 76326-252 CA76501-568 76326-232 76326-224 76326-246 CA76501-566 CA76501-588 76326-226 CA76501-596 76326-248 CA76501-586 76326-258 CA76501-594 76326-216 76326-238 76326-218
Megafuge™ ST4 Plus-MD Centrifuges, Thermo Scientific
Centrifuges Benchtop Centrifuges
Ideal for routine processing, these centrifuges combine exceptional capacity, ergonomics, and value with market-leading capabilities including unparalleled productivity and throughput, as well as exceptional ergonomics.

  • Patented lid locking technology
  • Easily adjustable speed, time, and temperature during the run
  • Simple operation with an intuitive control panel and digital display of parameters
  • Space-saving design conserves workspace, while ergonomic height ensures comfortable sample handling
  • Energy efficient design saves up to 40% of energy on industry standard protocols

The Megafuge™ ST4 Plus & Pro series can process up to 40×50 ml conical tubes, 196×5/7 ml blood tubes, 24 micro plates. The Megafuge™ ST4 Plus & Pro general-purpose centrifuge series can process clinical protocols, cell culture applications, and microplate processing. The refrigerated version (Megafuge™ ST4R Plus) is designed for handling temperature-sensitive sample processing between –10 and +40 °C.

Auto-Lock™ III Rotor System allows easy push-button installation and exchange of rotors, as well as easy access to the rotor chamber for quick cleaning.

ClickSeal™ Bucket Sealing System seals with a snap, eliminating screws and complicated clips. It is glove-friendly and certified for Biocontainment by CAMR in Porton Down, UK.

SMARTSpin™ Technology is an advanced rotor management system that maximizes acceleration, braking, and residual load imbalances for each rotor and bucket option. The system optimizes safety and improves separations.

Certifications: Certifications EN/UL/IEC 61010-1, EN/UL/IEC 61010-2-020 and 61010-2-101, EN/UL/IEC 61326-1, EN/UL/IEC 61326-2-6, FCC part 15.

Delivery information: All packages include: TX-1000 rotor cross, TX-1000 buckets (set of 4), ClickSeal biocontainment lids (set of 4).
Cell culture package includes 50 ml adapter (set of 4), 15 ml adapter (set of 4).
Blood tube package includes: 5/7 ml adapter (set of 4), 10 ml adapter (set of 4).

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty