Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TX-750 Swinging Bucket Rotor Packages, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

75016109 75016110
76326-280EA 7635.1 USD InStock
76326-280 76326-390
TX-750 Swinging Bucket Rotor Packages, Thermo Scientific
Centrifuges Rotors
Increase lab productivity with the Thermo Scientific™ TX-750 4×750ml Swinging Bucket Rotor packages, delivering maximum flexibility at 4700 rpm.

Auto-Lock™ III Rotor System allows easy push-button installation and exchange of rotors, as well as easy access to the rotor chamber for quick cleaning.

Delivery information: All Packages Include: TX-750 rotor cross, TX-750 buckets (set of 4), ClickSeal biocontainment lids (set of 4)
Cell culture package includes 50 ml adapter (set of 4), 15 ml adapter (set of 4)
Blood tube package includes: 5/7 ml adapter (set of 4), 10 ml adapter (set of 4)

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty