Would you like to log in again?
Biohazard Spill Kit, Bucket, Spill Magic, Acme United
Supplier: Acme United
97501
76327-120CS 559.7 USD InStock
76327-120
Biohazard Spill Kit, Bucket, Spill Magic, Acme United
Spill Control Materials
Spill magic single use biohazard cleanup kit safely removes blood and bodily fluids.
  • 12 oz. bag of all-purpose spill clean up to quickly absorb and remove spill
  • Personal protective equipment components of the kit help minimize exposure of personnel to potentially harmful substances
  • Safely clean up potentially infectious bodily fluids such as vomit, spit, blood, or urine with tools and disposal bags
  • Disposable single use kit to prevent crosscontamination
The disposable kit includes 12 oz. Spill Magic Absorbent, plastic bag, goggles, gloves, brush and dustpan, chemical surface wipe, anti-microbial towelette and I.D. tag.
Accessories information: Tools needed to clean up spill included.
Caution: Used Spill Magic must be disposed of in accordance with any federal, state or local laws and regulations governing the absorbed liquid or material. Do not use to absorb hydrofluoric acid. Do not pour down drain.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty