Would you like to log in again?
All-Purpose Spill Clean Up Shaker, Spill Magic, Acme United
Supplier: Acme United
97504
76327-126CS 108.08 USD InStock
76327-126
All-Purpose Spill Clean Up Shaker, Spill Magic, Acme United
Spill Control Materials
Spill magic lightweight, high performance, all-purpose absorbent powder removes a liquid or semi-liquid spill of any source or viscosity almost instantly.
  • 25 oz. shaker of lemon scented absorbent‚powder
  • Shaker for easy dispersal of powder on spill
  • Quick and easy to use
  • Non-toxic
  • Absorbs spill instantly leaving the surface clean & dry
  • For any spill (except Hydrofluoric Acid)
  • For use on asphalt, tile or concrete surfaces
Spill magic is a quick clean up solution that leaves the surface dry and residue free. Save time, money and reduce slip and fall instances.
The powder is ideal for cleaning industrial, food or biohazard spills. The portable shaker bottle allows for easy dispersal of the powder around the spill.
Accessories information: Accessories not included.
Caution: Used Spill Magic must be disposed of in accordance with any federal, state or local laws and regulations governing the absorbed liquid or material. Do not use to absorb hydrofluoric acid. Do not pour down drain.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty