Would you like to log in again?
Acid Neutralizer Spill Clean Up Bottle, Spill Magic, Acme United
Supplier: Acme United
SM202DBAN
76327-190CS 201.56 USD InStock
76327-190
Acid Neutralizer Spill Clean Up Bottle, Spill Magic, Acme United
Spill Control Materials
Spill magic lightweight, high performance acid neutralizing absorbent powder removes any liquid or semi-liquid spill situation instantly.
  • 5 lbs. bottle of acid neutralizing absorbent powder
  • Quick and easy to use
  • Non-toxic
  • Absorbs spill instantly leaving the surface clean & dry
  • For any spill (except Hydrofluoric Acid)
  • For use on asphalt and concrete surfaces
Leaves the surface clean and dry - no spill residue. Save time, money and reduce slip and fall instances. The powder is ideal for cleaning industrial or acidic spills.
Accessories information: Accessories not included.
Caution: Used spill magic must be disposed of in accordance with any federal, state or local laws and regulations governing the absorbed liquid or material. Do not use to absorb hydrofluoric acid. Do not pour down drain.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty