Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® LTS Compatible Pipette Tips

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
63305058 63305053 63305059 63305067 63305062 63305076 63305072 63305056 63305065 63305070 63305068 63305061 63305057 63305055 63305060 63305064 63305063 63305074 63305069 63305066 63305075 63305073 63305077 63305071 63305052
76327-212CS 294.14 USD InStock
76327-212 76322-178 76322-530 76323-938 76322-158 76322-532 76327-210 76323-936 76322-534 76323-934 76323-932 76322-172 76322-174 76322-176 76323-942 76323-940 76327-220 76323-946 76323-944 76322-180 76322-160 76322-162 76322-164 76327-218 76327-216
VWR® LTS Compatible Pipette Tips
Pipette Tips
VWR® LTS™ Compatible pipette tips feature a flexible seal area and a positive stop resulting in a secure seal with light ejection force on LTS™ compatible pipettors.

  • Compatible with the LTS™ style pipettors
  • Maximizes sample recovery with low binding resin and a fine point design
  • Improves ergonomics by reducing the force required to attach and eject a tip
  • Autoclavable at 15 psi, 122 °C for 15 minutes
  • Manufactured under an ISO 9001 quality management system

VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropylene pipette tips, enabling viscous samples like protein and DNA solutions to dispense easily without collecting inside the tip.

Certifications: Certified RNase-, DNase-, endotoxin-, metal-, and human DNA-free.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty