Would you like to log in again?
ACA CLEANERT 200 MG 6 ML PK30
Supplier: Agela Technologies
Cleanert ACA 200mg/6ml^ Cleanert ACA, Solid phase extraction of Acrylamide from drinking water is shown with excellent recoveries and accuracy (RSD<5) using Cleanert ACA, Size: 200mg/6mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ACA CLEANERT 200 MG 6 ML PK30 76327-252 ACA2006 Each Retrieving
Pack of 30 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty