Would you like to log in again?
COLUMN FLASH HILIC 40-60UM 100A 120G PK5
Supplier: Agela Technologies
Claricep* Screw-on Flash HILIC column 120g^ Claricep* Screw-on Flash HILIC column, Spherical, C-Series, Convenience of sample loading and adaptability, Average particle size: 40-60um, Average pore size: 100A, Silica amount: 120g
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN FLASH HILIC 40-60UM 100A 120G PK5 76327-400 C-SH240120-0 Each Retrieving
Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty