Would you like to log in again?
CARTRIDGE DURSHL C8L 150A 10UM 21.2X10MM
Supplier: Agela Technologies
Durashell C18 (L) Guard Cartridge, Pore size: 150A, Particle size: 10um, Dimension: 10 x 21.2mm^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARTRIDGE DURSHL C8L 150A 10UM 21.2X10MM 76327-468 DC900120-L Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty