Would you like to log in again?
COLUMN INNOVAL SILICA 5UM 100A 150X3.9MM
Supplier: Agela Technologies
Innoval Silica LC Column, Particle size: 5um, Pore size: 100A, Dimension: 150 x 3.9mm^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN INNOVAL SILICA 5UM 100A 150X3.9MM 76327-670 ISI9515039-0 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty