Would you like to log in again?
COLUMN FLASH SPHERICAL NH2 40-60UM 330G
Supplier: Agela Technologies
Flash Spherical NH2 Column, Particle size: 40-60um, Pore size: 100A, Silica amount: 330g^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN FLASH SPHERICAL NH2 40-60UM 330G 76328-038 SN240330-0 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty