Would you like to log in again?
FRIT PE 10UM 1/16 20G FLASH COLUMNPK20
Supplier: Agela Technologies
Flash Frit 20g, Bottom Frit, Polyethylene, Particle size: 10um, 1/16in, For screw-on Flash column^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FRIT PE 10UM 1/16 20G FLASH COLUMNPK20 76328-834 FCS020-SX Each Retrieving
Pack of 20 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty