Would you like to log in again?
FRIT FLASH COLUMN 1/8 TOP 40G 20UM PK20
Supplier: Agela Technologies
Flash Frit 40g, top Frit, Polyethylene, Particle size: 20um, 1/8in, For screw-on Flash column^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FRIT FLASH COLUMN 1/8 TOP 40G 20UM PK20 76328-836 FCS040-SS Each Retrieving
Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty