Would you like to log in again?
COLUMN PROMOSIL NH2 5UM NH2 100A
Supplier: Agela Technologies
Promosil NH2 column, Pore size: 100 armstrong, Particle size: 5um, Dimension: 300 x 4.0mm^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN PROMOSIL NH2 5UM NH2 100A 76329-094 PM753004-0 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty