Would you like to log in again?
COLUMN DURASHELL RP 20-35UM 80G PK5
Supplier: Agela Technologies
Flash Spherical Durashell RP Column, Particle size: 20-35um, Pore size: 100A, Silica amount: 80g^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN DURASHELL RP 20-35UM 80G PK5 76329-178 SDR230080-0 Each Retrieving
Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty