Would you like to log in again?
VWR CARBOY EZBIO ST AMB PP 83MM CAP 10L
Supplier: VWR International
VWR Carboy Ezbio St Amb Pp 83Mm Cap 10L^ VWR EZBio Carboy, Dark Amber PP, 10L, No Spigot, VersaCap 83mm, Double Bagged, Gamma Sterilized, maximizes storage efficiency and ease of use, distinctive rectangular shape saves valuable bench space, Larger handle
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR CARBOY EZBIO ST AMB PP 83MM CAP 10L 76329-428 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty