Would you like to log in again?
SMARTSCAN FAST/SECURE DATA RECOGNITION
Supplier: Mettler Toledo
SMARTSCAN COMPATIBLE with ANALYTICAL^ SmartScan, Fast and Secure Data Recognition, RFID module for direct integration into titrator systems with InMotion autosamplers for XPR analytical balances, Securely transfer titration sample information from the bal
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SMARTSCAN FAST/SECURE DATA RECOGNITION 76329-442 30459915 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty