Would you like to log in again?
Clarity Drugs of Abuse Urine Test Cups, Clarity Diagnostics
Supplier: Clarity Diagnostics
CD-CDOA-7125 CD-CDOA-7115 CD-CDOA-6124 CD-CDOA-6125
76329-462BX 239.5 USD InStock
76329-462 76329-460 76329-456 76329-459
Clarity Drugs of Abuse Urine Test Cups, Clarity Diagnostics
Drug Tests
Integrated drug panel and testing cup in one.
  • CLIA Waived and CLIA Moderate custom configurations available
  • Simple one-step procedure
  • Automatic split-specimen with no urine contamination
  • Screw lid for a no-leak cup
Test for: AMP, BAR, BUP, BZD, COC, mAMP, MDMA, MTD, OPI, OXY, PCP, PPX, TCA, and THC in urine with or without adulteration parameter.
Adulteration kit supplied as a kit component and detects variance in pH, Specific Gravity, Oxidants, PCC, Creatinine, Nitrites, and Glutaraldehyde for error proof testing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty