Would you like to log in again?
VWR CARBOY EZBIO HDPE 83MM VERSACAP 5L
Supplier: VWR International
VWR Carboy Ezbio Hdpe 83Mm Versacap St 5L^ VWR EZBio Carboy, HDPE, 5L, No Spigot, VersaCap 83mm, Double Bagged, Gamma Sterilized, maximizes storage efficiency and ease of use, distinctive rectangular shape saves valuable bench space, Larger handles on top,
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR CARBOY EZBIO HDPE 83MM VERSACAP 5L 76329-468 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty