Would you like to log in again?
VWR® Graduated Wide Mouth AC Round Bottles, Clear
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
293143 VWR89044-098 293145 293110 293144 293128 293121 293114 293116 293129 293107 293103 293135 293136 293146 295127 293122
76329-562CS 407.9 USD InStock
76329-562 76329-570 10862-318 10862-328 10862-376 10862-366 10862-364 76355-682 76329-554 76329-630 76329-564 76329-574 76329-548 76329-558 76329-546 10862-370 76329-556
VWR® Graduated Wide Mouth AC Round Bottles, Clear
Bottles Glass Bottles
VWR® Clear, graduated, wide mouth AC round bottles are manufactured from soda-lime glass that conforms to USP Type III requirements.
  • Soda-lime glass fabrication
  • Graduated
  • Wide mouth
Bottles are available in three packaging options. They may be purchased in convenience packs with closures attached to help maintain cleanliness, in bulk packs with unattached closures in separate bags, or bulk packed without closures.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty