Would you like to log in again?
GLOVES MULTI-LAYER BLACK SZ 9 20MIL 14IN
Supplier: NEWELL BRANDS INC
Glove rolled cuff 20 mil 14in size 9^ Glove, 20 mil, 14in, Multi-Layer black butyl, unsupported, rolled cuff, Z-grip, The only Butyl glove on the market with an exclusive embossed exterior finish, meaning slippery objects can be handled without risk, size
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES MULTI-LAYER BLACK SZ 9 20MIL 14IN 76329-580 34651209 Case of 6 (1/Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty