Would you like to log in again?
DISH ATTACHMENT AS 501.5 W/VESSEL 1.120G
Supplier: IKA WORKS
AS 501.5 Dish attachment^ AS 501.5 Dish attachment - For smoothly shaking dishes and smooth mixing in vessels with a large, flat bottom. A plastic foil with mild adhesive prevents the vessel from slipping. Dimensions (W x D x H): 450 x 450 x 45 mm, Set-up
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DISH ATTACHMENT AS 501.5 W/VESSEL 1.120G 76329-682 0002339600 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty