Would you like to log in again?
GRINDING CHAMBER MT 40.10 ST 40 ML PK10
Supplier: IKA WORKS
MT 40.10 sterile^ MT 40.10 sterile, Disposable grinding chamber with a stainless steel beater reduces soft, medium, hard and brittle materials with a Mohs hardness of up to 5 The chamber is made of transparent plastic so grinding tests can be observed at
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GRINDING CHAMBER MT 40.10 ST 40 ML PK10 76329-790 0020001984 Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty