Would you like to log in again?
IKA PETTE fix and vario, Single Channel Pipettes, IKA WORKS
Supplier: IKA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
0020011223 0020011210 0020011225 0020011228 0020011218 0020011226 0020011216 0020011224 0020011221 0020011213 0020011219 0020011227 0020011214 0020011222 0020011211 0020011215 0020011220 0020011217
76329-760EA 224.57 USD InStock
76329-760 76329-770 76329-804 76329-802 76329-808 76329-806 76329-764 76329-774 76329-796 76329-762 76329-794 76329-772 76329-768 76329-800 76329-766 76329-810 76329-776 76329-798
IKA PETTE fix and vario, Single Channel Pipettes, IKA WORKS
Pipettes Micropipettes
An ergonomic feel combined with uncompromising precise performance: the IKA single fix and vario pipettes can be used for many applications in the modern laboratory.
  • Exchangeable grips in different shapes and materials
  • Tip cone and piston are coated with a high quality diamind-like carbon (DLC)
  • Large display which shows set volumes in easily read characters
  • User-friendly IKA software
  • Simple maintenance and adjustment
Color coding simplifies the rapid selection of a suitable pipette. Resistance to impact, UV and chemicals ensures that the tip cone provides long-term reliable dosing. The PETTE fix and vario are fully autoclavable, therefore ensuring easy sterilization. This guarantees fault-free, precise and safe operation.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty