Would you like to log in again?
MILLING TUBE MMT 100.1 0.2816KG WT 100ML
Supplier: IKA WORKS
MMT 100.1 100ml^ MMT 100.1, Stainless steel milling tubes that can be used and cleaned in a dishwasher multiple times. The package includes a set of spare parts so that wearing parts can be replaced if necessary. Useful volume: 100 ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MILLING TUBE MMT 100.1 0.2816KG WT 100ML 76329-816 0020016315 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty