Would you like to log in again?
Hula Dancer basic Orbital Shaker, IKA
Supplier: IKA
0025003634 0025004320
76329-818EA 633.14 USD InStock
76329-818 76329-686
Hula Dancer basic Orbital Shaker, IKA
Shakers and Mixers Orbital Shakers
The Hula Dancers are powerful orbital shakers.
  • Robust housing
  • High stability
  • Touch function
Hula Dancer basic has a shaking diameter of 4.5 mm and allows a maximum load of 100 g. Speed is fixed at 2800 rpm.
Permissible relative humidity is 80%.
Certifications: IP protection class according to DIN EN 60529: IP 42.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty