Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Orbital Shaker, KS 501 digital

Supplier: IKA
0025002986
76329-906EA 5812.6 USD InStock
76329-906
Orbital Shaker, KS 501 digital
Shakers and Mixers Orbital Shakers
Low profile laboratory shaker with digital display which has a large mounting surface. Continuous operation is guaranteed even under extreme loads. Ideal for larger vessels of 250 ml and over, e.g. round bottomed flasks, Erlenmeyer flasks, culture flasks and bottles. Variety of different vessels can be accommodated by choosing from the range of optional holding attachments (not included with shaker).

  • Infinitely variable speed control continuous operation or timer operation from 0 to 55 min
  • Can be used at 5 to 50 °C (80% relative humidity), in an incubator or cold room
  • Integrated USB and RS232 interface

Certifications: IP protection class according to DIN EN 60529: IP 21.

Accessories information: Shaker KS 501 digital plus accommodates a maximum of 50×25, 48×50, 25×100, 16×250 or 12×500 ml flasks.

Ordering information: Supplied without attachments, required accessories must be ordered separately.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty