Would you like to log in again?
OVERHEAD STIRRER EUROSTAR 400 CONTROL
Supplier: IKA WORKS
Powerful laboratory stirrer^ Powerful laboratory stirrer for high viscous applications and intensive mixing for quantities up to 150 l (H2O). The control stirrer comes equipped with a RS 232 and a USB interface to control and document all parameters
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
OVERHEAD STIRRER EUROSTAR 400 CONTROL 76329-924 0025000757 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty