Would you like to log in again?
VACUUM CONTROLLER VC 10 50/60HZ 1000 MA
Supplier: IKA WORKS
VC 10 Vacuum controller^ VC 10 Universal vacuum controller for a two-point calibration of multiple vacuum sources. High-resolution TFT display ensures intuitive operation and navigation, Vacuum sensor type: ceramic Al2O3, Voltage: 100 - 240 V, Dimensions:
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VACUUM CONTROLLER VC 10 50/60HZ 1000 MA 76329-956 0020005132 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty