Would you like to log in again?
SEPARATING FUNNEL ATTACHMENT AS 501.3
Supplier: IKA WORKS
AS 501.3 Separating funnel attachment^ AS 501.3 Separating funnel attachment - For shaking out, eluting, extracting, gassing out, dissolving, Dimensions (W x D x H): 480 x 505 x 190 mm, Capacity: 4 x 500 or 3 x 1000 or 2 x 2000 ml, Weight: 3720 g
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SEPARATING FUNNEL ATTACHMENT AS 501.3 76329-958 0008000400 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty