Would you like to log in again?
GLOVES PERFECT FIT CUT RESIST S DZ 12 PR
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
SPECTRA GLOVE CUT RESISTANT SMALL^ Spectra* Seamless Knit Glove, Perfect Fit, Cut Resistant, 10 gauge medium weight, CE Cut level 4, abrasion level 3, Power cuff colors, launderable, Size: Small
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES PERFECT FIT CUT RESIST S DZ 12 PR 76330-018 PF10-GY-S Dozen(1/Itm/Bx) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty