Would you like to log in again?
Hypersil™ HS C18 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
Hypersil™
41010-019323B 41003-252130 41005-152130 37108-254630 41010-159070A 41003-104630 41010-159270A 41010-059270A 41005-159570XL 41003-154630 41005-059070A 41005-259270A 41010-029323A 41005-052130 41010-018823A 41005-259270XL 41010-029323B 41005-259370XL 41003-254630 41005-259570XL 41010-259570A 41005-154630 41005-102130 41010-159370XL 41010-259070A 41010-109270A 41010-159270XL 41005-159370XL 41005-259370A 41010-059270XL 41005-159070A 41010-019023A 41005-159270A 41010-259270A 41010-259570XL 41005-054630 41005-252130 41005-104630 41003-102130 41005-109270XL 41005-159370A 41003-152130 41005-059270A 37105-254630 41005-254630 41005-159270XL 41005-159570A 41010-109270XL 41005-059270XL 41010-259270XL 41003-052130 41005-109270A 41003-054630 41010-259370XL 41010-259370A 41005-109070A 41005-259570A 41010-019323A 41010-159570XL 41010-059070A 41010-159370A 41010-109070A 41005-259070A 41010-159570A
76330-194EA 3961.73 USD InStock
76330-194 76330-230 76330-196 76330-232 76330-550 76330-198 76330-552 76330-554 76330-556 76330-558 76330-190 76330-192 76330-560 76330-562 76330-564 76330-566 76330-202 76330-568 76330-128 76330-204 76330-206 76330-208 76330-172 76330-570 76330-130 76330-174 76330-210 76330-572 76330-176 76330-212 76330-178 76330-574 76330-214 76330-576 76330-216 76330-578 76330-218 76330-184 76330-580 76330-186 76330-582 76330-584 76330-188 76330-220 76330-542 10046-208 76330-586 76330-222 76330-544 76330-588 76330-224 76330-546 76330-226 76330-548 76330-228 10046-216 76330-180 76330-182
Hypersil™ HS C18 HPLC Columns, Thermo Scientific
Chromatography Columns
Specifically developed for pharmaceutical analysis and purification.
  • Ideal for LC/MS applications - No detectable bleed (as independently tested)
  • Highly stable to ensure excellent run-to-run and lot-to-lot reproducibility and long column life
  • Scalable from analytical to prep
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty