Would you like to log in again?
COLUMN PREP HS HYPERSIL C18 10UM 50X250
Supplier: Thermo Scientific
Hypersil* HPLC Column 250 mm^ Hypersil* HPLC Column, preparative HS C18, Standard, Reversed Phase, Particle Size: 10 um, Pore Size: 100A, Max. Pressure: 2000 psi (140 bar), pH: 2 to 9, Surface Area: 350 m2/g, Temperature: 60deg C, USP Type: L1, Stationary Phase: C18, Dimensions: 250X50mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN PREP HS HYPERSIL C18 10UM 50X250 76330-198 41010-259570A Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty