Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hypersil™ PREP HS Si HPLC Columns

Supplier: Thermo Scientific
Hypersil™
41405-059070A 41405-152130 41405-054630 41405-102130 41405-159370XL 41405-059270XL 41405-259070A 41405-059270A 41405-259570XL 41405-019323B 41405-018823A 41405-029323B 41405-259370XL 41405-109270XL 41405-109270A 41405-259270A 41405-159270A 41405-104630 41405-254630 41405-052130 41405-159570A 41405-159570XL 41405-252130 41405-259370A 41405-159070A 41405-154630 41405-159270XL 41405-019023A 41405-019323A 41405-259270XL 41405-109070A 41405-159370A 41405-259570A 41405-029323A
76330-494EA 495.83 USD InStock
76330-494 76330-350 76330-496 76330-352 76330-330 76330-332 76330-310 76330-354 76330-356 76330-334 76330-312 76330-314 76330-358 76330-336 76330-316 76330-338 76330-318 76330-382 76330-360 76330-384 76330-340 76330-362 76330-386 76330-342 76330-320 76330-322 76330-324 76330-346 76330-304 76330-326 76330-348 76330-306 76330-328 76330-308
Hypersil™ PREP HS Si HPLC Columns
Chromatography Columns
Maximize retention and loading using Thermo Scientific™ Hypersil™ PREP HS Si preparative HPLC columns.

  • Silica to separate nonpolar and moderately polar organic compounds
  • USP L3 - high surface area particle with 100 Å pore size
  • pH stable from 2 to 7
  • Maximum temperature of 60 °C

Packed with a bare silica, the high surface area particles with a 100 Å pore size are made for normal phase chromatography of nonpolar and moderately polar organic compounds. Our preparative HPLC columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed. Preparative HPLC Column Hardware Options - based on column inner diameter (Int. Ø.) and column performance needs.

Standard threaded and standard flange hardware
Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use
Standard threaded hardware for column Int. Ø. <40 mm
Standard flange hardware for column Int. Ø. ≥40 mm
Standard hardware can be modified on request to be compatible with SFC

XtendedLife hardware
Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly
Compatible with SFC and is available in dimensions up to 50 mm Int. Ø.

Preparative guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty