Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hypersil™ PREP Phenyl HS HPLC Columns

Supplier: Thermo Scientific
Hypersil™
41505-259570XL 41505-054630 41505-102130 41505-109270A 41505-159370A 41505-159070A 41505-109270XL 41505-019023A 41505-254630 41505-159270XL 41505-029323A 41505-159570A 41505-259370XL 41505-259370A 41505-019323A 41505-104630 41505-029323B 41505-159370XL 41505-052130 41505-019323B 41505-018823A 41505-059270XL 41505-159570XL 41505-059270A 41505-252130 41505-259070A 41505-154630 41505-109070A 41505-059070A 41505-259270XL 41505-259270A 41505-159270A 41505-259570A 41505-152130
76330-392EA 693.92 USD InStock
76330-392 76330-394 76330-396 76330-498 76330-410 76330-398 76330-412 76330-510 76330-414 76330-512 76330-416 76330-514 76330-418 76330-516 76330-508 76330-408 76330-390 76330-388 76330-520 76330-420 76330-522 76330-500 76330-422 76330-400 76330-524 76330-502 76330-402 76330-526 76330-504 76330-404 76330-506 76330-528 76330-406 76330-518
Hypersil™ PREP Phenyl HS HPLC Columns
Chromatography Columns
Maximum retention and loading of aromatic and moderately polar compounds with Thermo Scientific™ Hypersil™ PREP HS Phenyl reversed phase preparative HPLC columns.

  • Enhanced selectivity for aromatic and moderately polar analytes
  • Selectivity complementary to that provided by alkyl chain phases
  • Moderate hydrophobicity compared to C18
  • Phenyl (USP L11) selectivity - high surface area particle with 100 Å pore size
  • pH stable from 2 to 8
  • Maximum temperature of 60 °C

A high surface area particle with a 100 Å pore size and a phenyl bonded phase gives the stationary phase moderate hydrophobicity, making it ideal for the separation of analyte mixtures with varying polarity and aromaticity perfect for small molecule and drug purification needs. Preparative HPLC Column Hardware Options - based on column inner diameter (int. Ø.) and column performance needs.

Standard threaded and standard flange hardware
Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use
Standard threaded hardware for column int. Ø. <40 mm
Standard flange hardware for column int. Ø. ≥40 mm
Standard hardware can be modified on request to be compatible with SFC

XtendedLife hardware
Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly
Compatible with SFC and is available in dimensions up to 50 mm int. Ø.

Preparative guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty