Would you like to log in again?
COLUMN LC RP MABPAC 4UM 50 X 1 MM
Supplier: Thermo Scientific
MAbPac* RP LC Column 1x50mm^ MAbPac* RP LC Column, Analytical, Particle Size: 4um, Pore Size: 1500A, Temperature: 110 deg C, Max. Pressure: 4,000 psi, pH: 1 to 7, packing Material: PS-DVB, Diameter (Metric)1.0mm, Dimensions: MABPAC RP ANALYTICAL 4uM 1.0X50MM
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN LC RP MABPAC 4UM 50 X 1 MM 76330-532 303182 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty