Specifically developed for pharmaceutical analysis and purification.
- Ideal for LC/MS applications - No detectable bleed (as independently tested)
- Highly stable to ensure excellent run-to-run and lot-to-lot reproducibility and long column life
- Scalable from analytical to prep