Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hypersil™ PREP BDS C18 HPLC Columns

Supplier: Thermo Scientific
Hypersil™
40010-259270A 40005-152130 40005-159270A 40005-109070A 40010-019323A 40005-259270A 40010-018823A 40010-259370A 40010-159570XL 40003-254630 40010-259570A 40010-259270XL 40010-059270XL 40010-059070A 40010-109070A 40005-059270XL 40003-102130 40005-109270XL 40005-059070A 40010-159270A 40010-019023A 40003-104630 40010-259570XL 40005-252130 40010-259370XL 40005-259370A 40005-259270XL 40010-029323A 40010-159270XL 40005-159070A 40005-159370A 40003-252130 40005-259570A 40003-054630 40005-102130 40005-054630 40010-159570A 40005-159270XL 40010-059270A 40010-109270A 40005-059270A 40010-159370XL 40003-052130 40003-152130 40010-159370A 40005-052130 40005-259370XL 40010-029323B 40005-159570XL 40005-254630 40010-259070A 40010-019323B 40005-109270A 40005-159570A 40005-104630 40010-159070A 40003-154630 40010-109270XL 40005-159370XL 40005-154630 40005-259070A 40005-259570XL
76330-590EA 4126.22 USD InStock
76330-590 76330-150 76330-152 76330-592 76330-154 76330-594 76330-630 76330-156 76330-596 76330-598 76330-158 76330-634 76330-636 76330-638 76330-628 76330-162 76330-164 76330-166 76330-168 76330-640 76330-642 76330-600 76330-644 76330-602 76330-646 76330-604 76330-648 76330-160 76330-650 76330-132 76330-652 76330-134 76330-654 76330-610 76330-136 76330-656 76330-612 76330-534 76330-138 76330-614 76330-658 76330-976 76330-616 76330-978 76330-606 76330-608 76330-170 76330-140 76330-142 76330-980 76330-144 76330-660 76330-146 76330-982 76330-662 76330-148 76330-664 76330-620 76330-622 76330-624 76330-626 76330-618
Hypersil™ PREP BDS C18 HPLC Columns
Chromatography Columns
Easily scale up your analytical method using Thermo Scientific™ Hypersil™ PREP BDS C18 reversed phase preparative HPLC columns.

  • C18 (USP L1) selectivity
  • Base deactivated silica
  • pH stable from 2 to 8
  • Maximum temperature of 60 °C

PREP columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed. These endcapped, silica-based columns offer C18 retention with base deactivation for reduced silanol activity and are an excellent choices for routine applications which require development from analytical to preparative LC scale. Preparative HPL column hardware options - based on column inner diameter (Int. Ø.) and column performance needs.

Standard threaded and standard flange hardware
Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use
Standard threaded hardware for column Int. Ø. <40 mm
Standard flange hardware for column Int. Ø. ≥40 mm
Standard hardware can be modified on request to be compatible with SFC

XtendedLife hardware
Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly
Compatible with SFC and is available in dimensions up to 50 mm Int. Ø.

Preparative guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty