Would you like to log in again?
COLUMN BDS HYPERSIL C18 5UM 50 X 25 MM
Supplier: Thermo Scientific
Hypersil* HPLC Column Prep BDS C18 25x50mm^ Hypersil* HPLC Column, Preparative BDS C18, Standard, Reversed Phase, Particle Size: 5um, Pore Size: 145A, Max. Pressure: 3500psi (240 bar, Ph: 2-8, Surface Area: 185m2/g, Carbon Load: 11%, Dimension: 25x50mm, for from analytical to preparative LC scale
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN BDS HYPERSIL C18 5UM 50 X 25 MM 76330-638 40005-059270XL Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty