Would you like to log in again?
Hypersil™ Preparative ODS C18 HPLC Columns
Supplier: Thermo Scientific
Hypersil™
40203-154630 40203-152130 40210-019323A 40203-104630 40205-259070A 40205-259370A 40210-019323B 40205-259570XL 40205-059270A 40210-259270A 40205-052130 40210-109070A 40205-109070A 40205-159270A 40205-059070A 40210-019023A 40205-259370XL 40205-159370A 40210-059270A 40210-109270A 40210-159270XL 40205-259570A 40203-252130 40205-159570A 40210-159370A 40210-059270XL 40203-254630 40205-109270A 40210-259370A 40205-259270A 40210-259570A 40205-054630 40210-259070A 40205-109270XL 40205-152130 40210-159070A 40210-159370XL 40205-102130 40210-159270A 40205-154630 40205-159070A 40205-159270XL 40205-259270XL 40210-259570XL 40210-059070A 40210-259270XL 40203-102130 40210-159570XL 40205-159370XL 40210-029323A 40203-054630 40210-109270XL 40205-159570XL 40210-159570A 40205-104630 40205-059270XL 40210-259370XL 40210-029323B 40205-252130 40205-254630 40203-052130 40210-018823A
76330-790EA 16309.26 USD InStock
76330-790 76330-792 76331-038 76330-794 76330-750 76330-830 76330-796 76330-752 76330-832 76330-754 76330-798 76330-834 76330-756 76330-836 76330-758 76330-826 76330-828 76330-760 76331-048 76330-762 76330-764 76331-046 76330-840 76331-044 76330-766 76330-768 76330-800 76331-040 76330-802 76330-838 76330-770 76330-772 76331-058 76331-056 76330-774 76330-810 76330-776 76331-054 76330-812 76330-778 76331-052 76330-814 76331-050 76330-804 76330-806 76330-808 76330-780 76330-782 76330-740 76330-784 76330-742 76330-786 76330-788 76330-744 76330-820 76330-746 76330-822 76330-748 76330-824 76330-816 76330-738 76330-818
Hypersil™ Preparative ODS C18 HPLC Columns
Chromatography Columns
Scale up your analytical method using Thermo Scientific™ Hypersil™ PREP ODS C18 reversed phase preparative HPLC columns. Our columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed. This C18 column is a general purpose column used in a broad range of applications, perfect for routine workflows.
  • C18 (USP L1) selectivity
  • pH stable from 2 to 8
  • Maximum temperature of 60 °C
  • Preparative HPLC Column Hardware Options - based on column inner diameter and column performance needs
Standard threaded and standard flange hardware: Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use. Standard threaded hardware for column Int.Ø <40 mm. Standard flange hardware for column int.Ø ≥40 mm. Standard hardware can be modified on request to be compatible with SFC.
XtendedLife Hardware: Improves column lifetime by incorporating an internal dynamic axial. Compression (DAC) system, ensuring the media bed is continuously packed tightly. Compatible with SFC and is available in dimensions up to int.Ø 50 mm. Preparative Guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.
Customer specific orders. If there is a Thermo Scientific stationary phase you are currently using and would like to obtain in a preparative scale not shown or would like to discuss a specific column dimension, we would be happy to discuss your requirements. Please contact your local sales representative for customer specific orders. Custom built columns are not eligible for returns.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty