Would you like to log in again?
Hypersil™ BDS Si HPLC Columns
Supplier: Thermo Scientific
Hypersil™
40310-029323B 40310-019323B
76330-914EA 606.73 USD InStock
76330-914 76330-912
Hypersil™ BDS Si HPLC Columns
Chromatography Columns
Hypersil™ PREP BDS Si preparative HPLC columns which are packed with a base deactivated silica for normal phase chromatography of nonpolar and moderately polar organic compounds.
These preparative HPLC columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed.
Normal phase chromatography: Base deactivated silica to separate nonpolar and moderately polar organic compounds, USP L3, pH stable from 2 to 7, maximum temperature of 60 °C.
Standard threaded and standard flange hardware:
Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use.
Standard threaded hardware for column int.Ø <40 mm
Standard flange hardware for column int.Ø ≥40 mm
Standard hardware can be modified on request to be compatible with SFC
XtendedLife hardware:
Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly
Compatible with SFC and is available in dimensions up to 50 mm int.Ø
Preparative Guards are a proven and cost effective way to extend the lifetime of your prep column. These preparative guard columns require the use of a preparative guard holder.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty