Would you like to log in again?
Hypersil™ PREP Si HPLC Columns
Supplier: Thermo Scientific
Hypersil™
40405-019323B 40405-259370XL 40405-159270XL 40405-059270A 40405-254630 40405-054630 40405-159270A 40405-059070A 40405-159570XL 40405-159370XL 40405-154630 40405-059270XL 40405-019023A 40405-159370A 40405-152130 40405-252130 40405-159070A 40405-109070A 40405-259270A 40405-259070A 40405-029323B 40405-019323A 40405-259570A 40405-102130 40405-104630 40405-109270A 40405-259370A 40405-052130 40405-159570A 40405-259570XL 40405-109270XL 40405-018823A 40405-259270XL 40405-029323A
76331-118EA 6731.95 USD InStock
76331-118 76331-116 76331-138 76331-114 76331-158 76331-136 76331-156 76331-112 76331-132 76331-154 76330-930 76331-152 76331-130 76330-932 76331-150 76330-934 76330-926 76330-928 76331-128 76331-148 76331-126 76331-146 76331-124 76331-144 76331-122 76330-920 76331-120 76331-142 76330-922 76331-140 76330-924 76330-936 76330-916 76330-918
Hypersil™ PREP Si HPLC Columns
Chromatography Columns
Hypersil™ PREP Si preparative HPLC columns scale up your normal phase chromatography analytical method. Our preparative HPLC columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed.
  • Normal phase chromatography:
  • Silica to separate nonpolar and moderately polar organic compounds
  • USP L3
  • pH stable from 2 to 7
  • Maximum temperature of 60 °C
Preparative HPLC column hardware options - based on column inner diameter (Int. Ø.) and column performance needs.
Standard threaded and standard flange hardware: Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use. Standard threaded hardware for column Int. Ø. <40 mm. Standard flange hardware for column Int. Ø. ≥40 mm. Standard hardware can be modified on request to be compatible with SFC.
XtendedLife hardware: Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly. Compatible with SFC and is available in dimensions up to 50 mm Int. Ø.
Preparative guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty