Would you like to log in again?
Hypersil™ Preparative BDS Phenyl HPLC Columns
Supplier: Thermo Scientific
Hypersil™
40505-019023A 40505-259370A 40505-029323B 40505-104630 40505-059070A 40505-059270A 40505-109070A 40505-259070A 40505-259370XL 40505-019323B 40505-159070A 40505-159370XL 40505-259570A 40505-159270XL 40505-102130 40505-254630 40505-159570A 40505-259270XL 40505-052130 40505-109270A 40505-059270XL 40505-259270A 40505-159270A 40505-252130 40505-259570XL 40505-109270XL 40505-152130 40505-018823A 40505-154630 40505-159370A 40505-029323A 40505-159570XL 40505-054630 40505-019323A
76330-970EA 21231.09 USD InStock
76330-970 76331-178 76330-972 76331-176 76330-974 76330-952 76331-174 76330-954 76331-172 76330-956 76330-948 76331-170 76331-190 76330-960 76331-168 76330-962 76330-940 76331-166 76331-188 76330-942 76330-964 76331-186 76331-164 76330-944 76330-966 76331-184 76331-162 76330-968 76330-946 76331-160 76331-182 76331-180 76330-958 76330-938
Hypersil™ Preparative BDS Phenyl HPLC Columns
Chromatography Columns
Maintain retention for aromatic and moderately polar analytes with Thermo Scientific™ Hypersil™ PREP BDS Phenyl reversed phase preparative HPLC columns as you scale up your method.
The stationary phase is base deactivated for reduced silanol activity and has moderate hydrophobicity, making it ideal for the separation of analyte mixtures with varying polarity and aromaticity.
The preparative HPLC columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed.
Standard threaded and standard flange hardware:
-Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use.
-Standard threaded hardware for column int.Ø <40 mm.
-Standard flange hardware for column int.Ø ≥40 mm.
-Standard hardware can be modified on request to be compatible with SFC.
XtendedLife hardware:
-Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly.
-Compatible with SFC and is available in dimensions up to int.Ø 50 mm.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty