Would you like to log in again?
Hypersil™ Preparative BDS C8 HPLC Columns
Supplier: Thermo Scientific
Hypersil™
40110-059270XL 40110-259070A 40105-259270A 40103-152130 40105-259370XL 40105-059270A 40105-259570A 40105-159370A 40110-259370XL 40110-019323B 40105-259070A 40105-059070A 40105-109270A 40103-154630 40103-054630 40110-259570A 40110-159370A 40105-104630 40110-259270A 40110-018823A 40110-059070A 40105-252130 40110-019323A 40105-052130 40110-159270A 40105-159270XL 40110-259270XL 40105-159570XL 40105-154630 40105-159570A 40103-254630 40110-159570XL 40110-159370XL 40103-104630 40105-259270XL 40105-159270A 40110-259370A 40103-102130 40105-109070A 40105-159070A 40110-159570A 40110-109270XL 40105-109270XL 40110-259570XL 40105-102130 40110-159270XL 40110-059270A 40105-159370XL 40110-109270A 40110-029323B 40110-109070A 40110-029323A 40105-059270XL 40110-159070A 40103-052130 40105-254630 40105-054630 40103-252130 40105-259570XL 40105-259370A 40110-019023A 40105-152130
76330-670EA 879.71 USD InStock
76330-670 76330-672 76330-990 76330-674 76330-992 76331-036 76330-676 76331-034 76330-994 76330-678 76330-710 76331-032 76331-030 76330-998 76330-712 76330-714 76330-706 76330-708 76331-008 76330-680 76331-006 76330-682 76331-004 76330-684 76330-720 76331-002 76330-686 76330-722 76331-000 76330-688 76330-724 76330-726 76330-716 76330-718 76330-690 76330-692 76331-018 76330-694 76331-016 76330-696 76331-014 76330-698 76331-012 76330-730 76330-732 76331-010 76330-734 76330-736 76330-728 76331-028 76331-026 76330-984 76331-024 76330-700 76330-986 76331-022 76330-666 76330-988 76330-702 76331-020 76330-668 76330-704
Hypersil™ Preparative BDS C8 HPLC Columns
Chromatography Columns
Scale up from analytical to a preparative LC method with Thermo Scientific™ Hypersil™ PREP BDS C8 reversed phase preparative HPLC column.
  • Successful retention and separation of moderately hydrophobic analytes
  • Ideal for fatty acids, which are too hydrophobic for analysis using C18 columns
  • Base deactivated silica
  • C8 (USP L7) selectivity
  • pH stable from 2 to 8
  • Maximum temperature of 60 °C
  • Higher analytical throughput compared to C18 alkyl chain chemistry
  • Quicker compound elution due to reduced hydrophobic interactions
These columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed. With similar selectivity to C18 columns and predictable elution order, C8 columns with base deactivation for reduced silanol activity are ideal for analytes with medium hydrophobicity or when a less hydrophobic phase is required to obtain optimum retention. The lower hydrophobicity allows compounds to elute more quickly, yielding faster separations.
Standard Threaded and Standard Flange Hardware; Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use. Standard threaded hardware for column int.Ø <40 mm. Standard flange hardware for column int.Ø. ≥40 mm. Standard hardware can be modified on request to be compatible with SFC.
XtendedLife Hardware; Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly. Compatible with SFC and is available in dimensions up to 50 mm int.Ø. Preparative Guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.
Preparative HPLC column hardware options - based on column inner Ø (int.Ø.) and column performance needs.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty