Would you like to log in again?
Hypersil™ PREP BDS Si HPLC Columns
Supplier: Thermo Scientific
Hypersil™
40305-109270XL 40305-052130 40310-029323A 40305-159370A 40305-159270A 40310-059070A 40303-254630 40305-159370XL 40305-259570A 40310-259570A 40305-152130 40305-104630 40303-102130 40310-159270XL 40303-104630 40310-059270A 40310-109270XL 40305-159070A 40305-059070A 40305-259070A 40305-054630 40310-259570XL 40305-259370A 40305-102130 40310-159370XL 40303-052130 40305-252130 40305-259270A 40310-019323A 40310-159570XL 40305-059270A 40310-019023A 40305-159570A 40310-259070A 40310-159370A 40310-159570A 40305-109070A 40310-259270A 40310-259370XL 40303-154630 40305-159270XL 40305-259370XL 40303-252130 40310-018823A 40310-059270XL 40310-159070A 40305-059270XL 40305-109270A 40305-259270XL 40310-109270A 40310-109070A 40310-159270A 40305-254630 40305-154630 40305-159570XL 40303-054630 40303-152130 40310-259370A 40310-259270XL 40305-259570XL
76330-870EA 173.93 USD InStock
76330-870 76330-872 76331-078 76331-110 76330-876 76331-076 76330-878 76330-910 76331-074 76331-072 76330-904 76331-070 76330-906 76330-908 76330-880 76330-882 76330-884 76331-088 76330-886 76330-842 76331-086 76330-844 76330-888 76331-084 76330-846 76331-082 76331-080 76330-890 76330-892 76330-850 76330-894 76330-896 76330-852 76330-898 76330-854 76331-098 76330-856 76331-096 76330-858 76331-094 76330-848 76331-092 76331-090 76331-108 76331-106 76331-104 76331-102 76330-860 76331-068 76331-100 76330-862 76331-066 76330-864 76330-900 76331-064 76330-866 76331-062 76330-902 76330-868 76331-060
Hypersil™ PREP BDS Si HPLC Columns
Chromatography Columns
Hypersil™ PREP BDS Si preparative HPLC columns which are packed with a base deactivated silica for normal phase chromatography of nonpolar and moderately polar organic compounds.
These preparative HPLC columns come in a variety of dimensions including an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed.
Normal phase chromatography: Base deactivated silica to separate nonpolar and moderately polar organic compounds, USP L3, pH stable from 2 to 7, maximum temperature of 60 °C.
Standard threaded and standard flange hardware:
Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use
Standard threaded hardware for column int.Ø <40 mm
Standard flange hardware for column int.Ø ≥40 mm
Standard hardware can be modified on request to be compatible with SFC
XtendedLife hardware:
Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly
Compatible with SFC and is available in dimensions up to 50 mm int.Ø
Preparative Guards are a proven and cost effective way to extend the lifetime of your prep column. These preparative guard columns require the use of a preparative guard holder.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty