Would you like to log in again?
CALIBRATION CERTIFICATE TEMP27/HUMID1 PT
Supplier: BINDER INC
Calibration Certificate Temp Kbf-S 1020^ Calibration certificate, temperature and humidity - temperature (according to DIN12880) and humidity measurement incl. Certificate, 27 temperature measuring points and 1 humidity measuring point, at 25 deg C / 60 % RH or at specified values, For KBF-S 1020
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CALIBRATION CERTIFICATE TEMP27/HUMID1 PT 76331-830 8012-0924 Each Retrieving
Temperature sensitive Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty